Learn More
Medchemexpress LLC 2-Aminotetralin | 2048491-06-9 | 98.06% | 269.36 | 50 MG

Supplier: Medchemexpress LLC HY16088750MG
FPT is a 2-Aminotetralin that acts as an efficacious partial agonist at 5-HT1A receptors and a full agonist at 5-HT1B and 5-HT1D receptors. It has demonstrated antiepileptic effects in Fmr1 knockout mice and exhibits anxiolytic-like and prosocial effects in various mouse models. This compound is used in research related to neurological conditions.
- Efficacious partial agonist at 5-HT1A receptors
- Full agonist at 5-HT1B and 5-HT1D receptors
- Weak agonist at 5-HT7R
- Antiepileptic effects in Fmr1 knockout mice
- Anxiolytic-like and prosocial effects in mouse models
- Purity of 98.06%
- Available in various sizes and forms
- Storage at 4°C, protected from light
- Shipping at room temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.