Learn More
Medchemexpress LLC 2-anilinobenzamide | 1211-19-4 | 99.1% | C13H12N2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13399810MG
Encompass_Preferred
SIRT-IN-3 is a small-molecule sirtuin inhibitor (CAS 1211-19-4) that inhibits SIRT1 (IC50 17 μM) and shows selectivity over SIRT2 and SIRT3. It is supplied as a high-purity research reagent suitable for biochemical and cell-based studies, available in solid form and as a DMSO solution, with defined storage recommendations.
Catalog No. 50-002-61846