Learn More
Selleck Chemical LLC 2-APQC E5907-1G
SDP
Supplier:  Selleck Chemical LLC E59071G
Encompass_Preferred
2-APQC is a potent activator of SIRT3 with a Kd value of 2 756 M It alleviates myocardial hypertrophy and fibrosis by regulating mitochondrial homeostasis exhibiting promising cardioprotective potential in vitro and in vivo It may serve as a potential candidate for future heart failure (HF) therapeutics
Catalog No. 50-000-42748