Learn More
Medchemexpress LLC 2'-azido-2'-deoxycytidine | 51034-68-5 | 98.8% | 268.23 g/mol | C9H12N6O4 | 10 MG

Supplier: Medchemexpress LLC HY15244810MG
2'-Azido-2'-deoxycytidine is an azidonucleoside research reagent and nucleoside analogue used in biochemical and oncology research. It contains an azide group for click-chemistry applications and requires phosphorylation to act as an inhibitor of nucleotide reductase. Supplied as a powder for laboratory use.
- Compatible with copper-catalyzed and strain-promoted azide-alkyne cycloaddition (CuAAC and SPAAC).
- Acts as a nucleoside analogue relevant to antitumor research.
- High purity (98.8%).
- Molecular formula C9H12N6O4; molecular weight 268.23 g/mol.
- Supplied as a powder with storage recommendations: -20°C (3 years) or 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.