Learn More
Medchemexpress LLC 2'-Azido-2'-deoxycytidine | 51034-68-5 | C9H12N6O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY152448
Encompass_Preferred
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue with broad antitumor activity, targeting indolent lymphoid malignancies. Its anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Functions as a click chemistry reagent due to its Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Requires double phosphorylation to become an inhibitor of nucleotide reductase
Catalog No. 50-003-74665