Learn More
Medchemexpress LLC 2'-Azido-2'-deoxycytidine | 51034-68-5 | C9H12N6O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1524485MG
Encompass_Preferred
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue that exhibits broad antitumor activity against indolent lymphoid malignancies by inhibiting DNA synthesis and inducing apoptosis. It functions as a click chemistry reagent due to its azide group, capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Purine nucleoside analogue
  • Broad antitumor activity
  • Functions as a click chemistry reagent
  • Participates in CuAAc and SPAAC reactions
  • Requires double phosphorylation to inhibit nucleotide reductase
Catalog No. 50-003-73552