Learn More
Medchemexpress LLC 2'-Azido-2'-deoxycytidine | 51034-68-5 | C9H12N6O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15244850MG
Encompass_Preferred
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue known for its broad antitumor activity against indolent lymphoid malignancies. It primarily works by inhibiting DNA synthesis and inducing apoptosis. The compound also functions as a click chemistry reagent, utilizing its Azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Purine nucleoside analogue with broad antitumor activity
  • Inhibits DNA synthesis and induces apoptosis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Requires double phosphorylation to inhibit nucleotide reductase
Catalog No. 50-003-73187