Learn More
Medchemexpress LLC 2'-Azido-2'-deoxycytidine | 51034-68-5 | MFCD00043047 | C9H12N6O4 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY152448200MG
Encompass_Preferred
2'-Azido-2'-deoxycytidine is a purine nucleoside analogue with broad antitumor activity, especially against indolent lymphoid malignancies. Its anticancer mechanisms include inhibiting DNA synthesis and inducing apoptosis. It also functions as a click chemistry reagent, capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. This azidonucleoside requires diphosphorylation to inhibit nucleotide reductase.
  • Broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis and induces apoptosis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Requires diphosphorylation for nucleotide reductase inhibition
Catalog No. 50-003-73888