Learn More
Apexbio Technology LLC 2'-Azido-dATP 10ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B79970.01
Encompass_Preferred
2p-Azido-dATP is a sugar-modified nucleoside triphosphate analog It incorporates a 2 -azido group substitution at the sugar moiety which allows its enzymatic integration into nucleic acids and leads to oligonucleotide derivatives with modified biochemical and biophysical characteristics 2p-Azido-dATP exerts its biological activity primarily through structural incorporation into nucleic acids during polymerization resulting in increased nuclease stability enhanced target binding affinity and reduced immunogenicity Based on these properties 2p-Azido-dATP holds research potential in nucleic acid therapeutics such as antisense oligonucleotides siRNA development gene-editing and aptamer-based applications
Catalog No. 50-000-87614