Learn More
Medchemexpress LLC 2-((Azido-PEG8-carbamoyl)methoxy)acetic acid | 846549-37-9 | 98.6% | C22H42N4O12 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140459
Encompass_Preferred
This PEG-based PROTAC linker, 2-((Azido-PEG8-carbamoyl)methoxy)acetic acid, is designed for the synthesis of PROTACs. It functions as a click chemistry reagent, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. It is intended for research use only.
  • Utilizes PEG-based PROTAC linker technology
  • Features an azide group for click chemistry reactions
  • Supports copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Suitable for PROTAC synthesis
Catalog No. 50-003-52889