Learn More
Medchemexpress LLC 2-Benzofurylboronic acid | 98437-24-2 | 161.95 | 10 G

Supplier: Medchemexpress LLC HYW00411610G
2-Benzofurylboronic acid is a chemical compound with the molecular formula C8H7BO3 and a molecular weight of 161.95. It appears as a white to off-white solid and has a high purity of 99.76% by HPLC.
- Molecular formula: C8H7BO3
- Molecular weight: 161.95
- Purity (HPLC): 99.76%
- Appearance: White to off-white solid
- Storage at -20°C in sealed storage, away from moisture
- Storage in solvent at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.