Learn More
eMolecules​ 1618666-29-7 | 2-Benzothiazolylzinc bromide, 0.50 M in THF | Synthonix279.460 | C7H4BrNSZn | 0.000 | [Br-].[Zn+]c1nc2ccccc2s1 | 25ml | 627353494
Supplier:  eMolecules​ 627353494
Encompass_Preferred
2-Benzothiazolylzinc bromide, 0.50 M in THF | Synthonix | 1618666-29-7279.460 | C7H4BrNSZn | 0.000 | [Br-].[Zn+]c1nc2ccccc2s1 | 25ml | 627353494
Catalog No. 50-269-5447