Learn More
Medchemexpress LLC 2-benzyl-2,5-diazaspiro[3.4]octane dihydrochloride | 1159823-70-7 | MFCD11501173 | 99.0% | 275.22 | C13H20Cl2N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY3080910MG
Encompass_Preferred
2-Benzyl-2,5-diazaspiro[3.4]octane dihydrochloride is a small-molecule chemical intermediate used in medicinal chemistry and organic synthesis for preparing biologically active compounds. It is supplied as a white to off-white solid in small research pack sizes and is intended for laboratory use under appropriate safety controls.
  • High purity (>99%).
  • White to off-white solid appearance.
  • Suitable as a synthetic building block for drug discovery.
  • Available in multiple small pack sizes (5-500 mg) for research use.
  • Storage: sealed, away from moisture; recommend refrigerated storage and low-temperature solvent storage.
Catalog No. 50-002-97590