Learn More
Medchemexpress LLC 2-benzyl-9-(1-(naphthalen-1-yl)ethyl)-1H-purin-6(9H)-one | 2988566-71-6 | 98.2% | C24H20N4O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13299310MG
Encompass_Preferred
Hcyb1 is a selective, orally active phosphodiesterase 2 (PDE2) inhibitor used for research. It potently inhibits PDE2A (IC50 = 0.57 μM) with strong selectivity versus other PDE family members, modulates intracellular cGMP and cAMP, and has shown neuroprotective and antidepressant-like effects in preclinical cellular and animal studies. Supplied as a high-purity solid with defined solubility and storage recommendations.
Catalog No. 50-002-58289