Learn More
Medchemexpress LLC 2-benzylbenzoic acid | 612-35-1 | MFCD00002484 | 99.0% | 212.25 g/mol | C14H12O2 | 1 ML

Supplier: Medchemexpress LLC HYW01429510MM/1ML
2-Benzylbenzoic acid (CAS 612-35-1) is a small-molecule carboxylic acid used as a building block in organic synthesis and medicinal chemistry research. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO, and is intended for research use only.
- Useful building block for compound synthesis.
- Available as solid or 10 mM solution in DMSO for convenience.
- High purity (≈99.0%) suitable for research applications.
- Molecular weight 212.25 g/mol and formula C14H12O2 support stoichiometric calculations.
- CAS number 612-35-1 enables unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.