Learn More
Medchemexpress LLC 2-(benzyloxy)-1-ethanamine | 38336-04-8 | MFCD00235180 | 99.8% | 151.21 | C9H13NO | 5 G

Supplier: Medchemexpress LLC HYW0178345G
2-(Benzyloxy)-1-ethanamine is an organic biochemical reagent (synonym: 2-(Benzyloxy)ethylamine) used as a building block in life-science research. It is a colorless to light-yellow liquid with defined molecular properties and high reported purity; follow product documentation and the SDS for safe handling.
- Primary amine building block for chemical synthesis and derivatization.
- High purity (~99.8%) suitable for research use.
- Molecular formula C9H13NO; molecular weight 151.21 g·mol-1.
- Physical state: liquid; colorless to light yellow.
- Common packaging sizes: 5 g-100 g; larger quantities available by quote.
- Intended for laboratory research; consult SDS for storage and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.