Learn More
Medchemexpress LLC 2-(benzyloxy)-4-fluorobenzaldehyde | 202857-89-4 | 99.8% | C14H11FO2 | 250 MG

Supplier: Medchemexpress LLC 250MGHY76084
2-(Benzyloxy)-4-fluorobenzaldehyde is a drug intermediate used for the synthesis of various active compounds. This product appears as a white to off-white solid and is identified as a laboratory chemical for the manufacture of substances.
- Appears as a white to off-white solid
- Suitable as a drug intermediate for various active compounds
- Recommended storage at 4°C, protected from light
- For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
- Purity of 99.8% (LCMS)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.