Learn More
Medchemexpress LLC 2-(Benzyloxy)-4-fluorobenzaldehyde | 202857-89-4 | C14H11FO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY760845G
Encompass_Preferred
2-(Benzyloxy)-4-fluorobenzaldehyde (HY-76084) is a chemical substance identified for laboratory uses and the manufacture of other substances. It presents as a solid and is classified with hazards including acute oral toxicity, skin and serious eye irritation, and potential respiratory tract irritation.
  • Identified for laboratory chemicals and substance manufacturing.
  • Presents as a solid.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
  • Requires handling with precautions to avoid inhalation and contact with eyes and skin.
  • Store in a cool, well-ventilated area, tightly sealed, and protected from direct sunlight and ignition sources.
  • Recommended storage at 4°C, protected from light.
  • For research use only by qualified and experienced scientists.
Catalog No. 50-004-34102