Learn More
Medchemexpress LLC 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoic acid | 2768-56-1 | 99.3% | 239.22 g/mol | C11H13NO5 | 25 G

Supplier: Medchemexpress LLC 25GHYW067360
2-(((Benzyloxy)carbonyl)amino)-3-hydroxypropanoic acid is a Cbz-protected serine derivative supplied as a white to off-white solid for research use. It is used as a protected amino acid building block in peptide synthesis and organic chemistry.
- High purity (99.3%).
- Molecular weight 239.22 g/mol.
- White to off-white solid physical form.
- Stable as a powder with long-term storage at -20°C.
- Available in multiple pack sizes, including 25 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.