Learn More
Medchemexpress LLC 2-(((Benzyloxy)carbonyl)amino)propanoic acid | 4132-86-9 | 99.97% | 223.23 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW012381100G
Encompass_Preferred
2-(((Benzyloxy)carbonyl)amino)propanoic acid is an alanine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • For research use only
  • Available in multiple languages for SDS documentation
  • Provided as a solid, white to off-white appearance
  • Stable under recommended storage conditions
Catalog No. 50-004-52963