Learn More
Medchemexpress LLC 2-bromo-1-(4-chlorophenyl)propan-1-one | 877-37-2 | MFCD00018687 | 98.0% | 247.52 | C9H8BrClO | 100 MG

Supplier: Medchemexpress LLC HYW045376100MG
2-bromo-4'-chloropropiophenone is an organic intermediate used as a precursor in the synthesis of pharmaceutical compounds.
- Precursor in the synthesis of pharmaceutical compounds.
- CAS number 877-37-2.
- Molecular formula C9H8BrClO, molecular weight 247.52 g/mol.
- Typical purity 98.0% (reported by suppliers).
- Available pack sizes: 100 mg, 250 mg, 500 mg, 1 g.
- Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.