Learn More
Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.1 g/mol | C9H11BrN2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW14133850MG
Encompass_Preferred
2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog used in biochemical and cell biology research for DNA labeling and proliferation assays. It incorporates into DNA in place of thymidine and is used to detect proliferating cells, study DNA synthesis, and prepare DNA-protein crosslinks. Store refrigerated and follow the SDS and COA for safe handling and purity confirmation.
  • Brominated nucleoside analog for DNA labeling.
  • Incorporates into DNA in place of thymidine.
  • Useful for cell proliferation and DNA synthesis assays.
  • Supplied as a stable solid suitable for laboratory use.
  • Store at 2-8 °C per supplier recommendations.
  • Provided with certificate of analysis for purity verification.
Catalog No. 50-002-82680