Learn More
Medchemexpress LLC 2-bromo-4,5-dimethoxybenzyl alcohol | 54370-00-2 | MFCD02093896 | 98.0% | 247.09 | C9H11BrO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1376861G
Encompass_Preferred
Pinaverium impurity 1 (2-bromo-4,5-dimethoxybenzyl alcohol; CAS 54370-00-2) is an analytical impurity of the pharmaceutical active pinaverium. Supplied for research and analytical use, it is intended for impurity profiling, method development, and quality control. Consult the Certificate of Analysis for lot-specific purity and storage conditions.
  • Analytical reference standard for impurity profiling.
  • Chemical formula C9H11BrO3 and molecular weight 247.09.
  • Suitable for method development and assay validation.
  • Packaged in small quantities for laboratory use.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-83475