Learn More
Medchemexpress LLC 2-bromo-4'-chloropropiophenone (2-bromo-1-(4-chlorophenyl)-1-propanone) | 877-37-2 | MFCD00018687 | >98.0% | 247.52 g/mol | C9H8BrClO | 250 MG

Supplier: Medchemexpress LLC 250MGHYW045376
2-Bromo-4'-chloropropiophenone (CAS 877-37-2) is a bromo-chlorinated propiophenone used as a chemical precursor in organic synthesis and research chemistry, supplied in small research pack sizes with accompanying safety and technical documentation.
- Precursor in organic synthesis and pharmaceutical intermediate production.
- Molecular formula C9H8BrClO.
- Molecular weight 247.52 g/mol.
- Typical listed purity >98% (refer to datasheet for analytical details).
- Available in small research pack sizes, for example 250 MG.
- Product documentation includes a datasheet and safety data sheet for handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.