Learn More
Medchemexpress LLC 2-Bromo-4'-fluoroacetophenone | 403-29-2 | 99.2% | 217.04 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW008598100G
Encompass_Preferred
2-Bromo-4'-fluoroacetophenone is a photoinducible DNA cleavage inducer. It generates fluorine-substituted phenyl radicals upon excitation and irradiation to cleave DNA. This compound exhibits concentration-dependent DNA cleaving activity.
  • Generates fluorine-substituted phenyl radicals upon excitation and irradiation.
  • Induces DNA cleavage, converting circular supercoiled DNA to circular relaxed and linear DNA forms.
  • Exhibits concentration-dependent DNA cleaving activity.
  • Applicable for cancer research.
  • Cleaves ΦX174 RFI supercoiled DNA.
Catalog No. 50-004-52155