Learn More
Medchemexpress LLC 2-Bromo-5-fluoropyridine | 41404-58-4 | 99.5% | 175.99 | 100 G
SDP
Supplier:  Medchemexpress LLC HY32748100G
Encompass_Preferred
2-Bromo-5-fluoropyridine is an intermediate used in organic synthesis. It can undergo Suzuki coupling reactions with boronic acid and is utilized in the synthesis of GSK1292263A, a GPR119 receptor agonist. This compound appears as a solid below 30°C and a liquid above 31°C, with a colorless to off-white appearance. It is soluble in DMSO and various in vivo protocols, offering flexibility for different research applications.
  • Intermediate for organic synthesis
  • Undergoes Suzuki coupling reactions
  • Used in synthesis of GPR119 receptor agonist
  • Solid below 30°C, liquid above 31°C
  • Colorless to off-white appearance
  • Soluble in DMSO and various in vivo protocols
Catalog No. 50-004-00412