Learn More
Medchemexpress LLC 2-Bromo-5-iodopyridine | 73290-22-9 | 283.89 | 25 G

Supplier: Medchemexpress LLC 25GHYW003027
2-Bromo-5-iodopyridine is a biochemical reagent primarily used in life science research. It serves as a versatile biological material or organic compound, intended strictly for research purposes.
- Solid, off-white to light yellow appearance
- Store at 4°C, protected from light
- When in solvent, store at -80°C for up to 6 months or -20°C for 1 month, protected from light
- Soluble in DMSO at 100 mg/mL (352.25 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.