Learn More
Medchemexpress LLC 2-Bromo-5-nitropyridine | 4487-59-6 | 203.00 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00785810G
Encompass_Preferred
2-Bromo-5-nitropyridine is a biochemical reagent used as a biological material or organic compound for life science related research.
  • Used as a biological material for research
  • Used as an organic compound for research
  • Suitable for life science research applications
  • Soluble in DMSO at 100 mg/mL (ultrasonic)
  • Functions as a biochemical assay reagent
Catalog No. 50-004-46790