Learn More
Medchemexpress LLC 2-bromo-6-methoxynaphthalene | 5111-65-9 | MFCD00004062 | 97.0% | 237.09 g/mol | C11H9BrO | 250mg
SDP
Supplier:  Medchemexpress LLC HYY0038R250MG
Encompass_Preferred
2-Bromo-6-methoxynaphthalene (Standard) is the analytical standard of 2-Bromo-6-methoxynaphthalene This product is intended for research and analytical applications 2-Bromo-6-methoxynaphthalene is an active intermediate in the production of anti-inflammatory agents like Naproxen and Nabumetone by Heck reaction 2-Bromo-6-methoxynaphthalene has potential anti-inflammatory properties and Tyrosine-protein inhibitor properties 2-Bromo-6-methoxynaphthalene can be used for the research of cancer[1]
Catalog No. 50-002-68964