Learn More
Medchemexpress LLC 2-bromo-6-nitrophenol | 13073-25-1 | MFCD02683216 | 98.0% | 218.01 g/mol | C6H4BrNO3 | 1 G

Supplier: Medchemexpress LLC HYW0408861G
2-bromo-6-nitrophenol is a brominated nitrophenol used as an intermediate in organic synthesis and research applications, commonly converted via 2-bromo-6-aminophenol to N-acetyl-2-bromo-6-aminophenol.
- Used as an intermediate in organic synthesis
- Purity 98.0%
- Molecular weight 218.01 g/mol
- Appearance: solid powder
- Storage: powder -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.