Learn More
Matrix Scientific 2-Bromodiphenylmethane, 23450-18-2, MFCD00079713, 1g
Supplier: Matrix Scientific 0057211G
Molecular Formula C13H11Br, Purity 98%, Molecular Weight 247.14, Boiling Point 192-198
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.