Learn More
Matrix Scientific 2-Bromofuran | ≥95% stabilized | 5g
Supplier:  Matrix Scientific 0760215G
Encompass_Preferred
Stabilization: Stabilized w/copper and layer of water. Hazard: Irritant/Light Sensitive/Refrigerate/Stored under Argon
Catalog No. 50-426-633