Learn More
Medchemexpress LLC 2-Butenamide, N-[1,1'-biphenyl]-4-yl-2-cyano-3-hydroxy- | 1173715-42-8 | 99.9% | 278.31 g/mol | C17H14N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1356585MG
Encompass_Preferred
hDHODH-IN-1 is a small-molecule inhibitor of human dihydroorotate dehydrogenase (hDHODH) with reported anti-inflammatory activity. It is supplied as a high-purity research reagent intended for in vitro and preclinical laboratory studies.
  • Potent inhibitor of human dihydroorotate dehydrogenase (hDHODH).
  • Demonstrated anti-inflammatory activity in biological assays.
  • High purity: 99.90% (LCMS).
  • Molecular formula C17H14N2O2 and molecular weight 278.31 g/mol.
  • Supplied as a 5 mg pack for laboratory research use.
  • Intended for research use only; not for human or clinical applications.
  • Certificate of analysis available for quality verification.
Catalog No. 50-002-22942