Learn More
Medchemexpress LLC 2-Butenedioic acid (2E)-, 1-ethyl ester, 4-[2-[[4-(phenylmethyl)-3-pyridinyl]carbonyl]hydrazide] | 1808089-27-1 | 99.3% | C19H19N3O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13187110MG
Encompass_Preferred
Ethyl LipotF is a small-molecule inhibitor of the fat mass and obesity-associated protein (FTO) used for research. It selectively inhibits FTO activity and increases N6-methyladenosine (m6A) levels in HeLa cells in a concentration-dependent manner. Supplied as a white to off-white solid with high reported purity for laboratory studies.
  • Selective FTO inhibitor suitable for cellular and biochemical assays.
  • Increases m6A levels in HeLa cells in a concentration-dependent manner.
  • High reported purity (>99%) supports reproducible results.
  • White to off-white solid form enables convenient storage and handling.
  • Applicable to studies of RNA methylation and epitranscriptomic regulation.
Catalog No. 50-002-60963