Learn More
Medchemexpress LLC 2-Carboethoxyimidazole | 33543-78-1 | 99.23% | 140.14 | 10 G

Supplier: Medchemexpress LLC HY2046010G
2-Carboethoxyimidazole is a biochemical reagent that can be utilized as a biological material or an organic compound for life science-related research. It is intended for research use only and is not sold to patients.
- Purity: 99.23%
- Appearance: solid
- Color: white to light brown
- Storage: powder: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months, -20°C for 1 month
- Solubility: DMSO: 125 mg/mL (891.97 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.