Learn More
Medchemexpress LLC 2-chloro-1-[2-[3-(trifluoromethyl)anilino]phenyl]ethanone | 2378626-29-8 | 99.7% | 313.70 g/mol | C15H11ClF3NO | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12526910MM 1ML
Encompass_Preferred
TED-347 is a potent, irreversible, covalent, allosteric inhibitor of the YAP-TEAD protein-protein interaction that blocks TEAD transcriptional activity and shows reported antitumor effects in preclinical studies. Reported biochemical activity includes an EC50 of 5.9 μM for TEAD4-Yap1 and a Ki of approximately 10.3 μM; the compound covalently modifies Cys-367 in TEAD4.
  • Irreversible covalent inhibitor of YAP-TEAD interaction.
  • Reported EC50 of 5.9 μM (TEAD4-Yap1) and Ki ≈ 10.3 μM.
  • Covalently modifies Cys-367 in TEAD4.
  • Demonstrated antitumor activity in preclinical studies.
  • Available as DMSO solution and as solid powder for formulation.
  • Molecular formula C15H11ClF3NO; molecular weight 313.70 g/mol.
  • High purity suitable for research applications (reported ≥98%).
Catalog No. 50-002-35941