Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | 99.9% | 129.55 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3283310G
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It also serves as an intermediate in the synthesis of other active compounds.
  • Biochemical reagent for life science research
  • Used in the synthesis of other active compounds
  • Identified as a halopyridinol disinfection by-product in drinking water samples
  • Demonstrates very low cytotoxicity to human hepatocellular carcinoma HepG2 cells in vitro
Catalog No. 50-004-01555