Learn More
Medchemexpress LLC 2-Chloro-3-pyridinol | 6636-78-8 | MFCD00006235 | 99.9% | 129.55 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY3283310MM/1ML
Encompass_Preferred
2-Chloro-3-pyridinol is a biochemical reagent used as a biological material or organic compound for life science research. It can also be utilized in the synthesis of other active compounds. Additionally, it is identified as a halopyridinol disinfection by-product found in drinking water samples. In vitro studies show that 2-Chloro-3-pyridinol exhibits very low cytotoxicity to human hepatocellular carcinoma HepG2 cells, with an EC50 > 10000 μM.
  • Biochemical reagent for life science research.
  • Used in the synthesis of other active compounds.
  • Identified as a halopyridinol disinfection by-product.
  • Exhibits very low cytotoxicity to HepG2 cells.
Catalog No. 50-004-04365