Learn More
Medchemexpress LLC 2-chloro-4,6-dimethoxy-1,3,5-triazine | 3140-73-6 | MFCD00075607 | 98.1% | 175.57 | C5H6ClN3O2 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0167181KG
Encompass_Preferred
Chlorodimethoxytriazine (2-chloro-4,6-dimethoxy-1,3,5-triazine) is a white to off-white solid used as a biochemical reagent and coupling agent in organic synthesis, particularly for peptide and amide bond formation. The compound has formula C5H6ClN3O2 and molecular weight 175.57, and is supplied with high assay purity for laboratory research.
  • High purity suitable for research applications.
  • Commonly used as a coupling reagent in peptide and amide synthesis.
  • Solid form facilitates storage and handling.
  • Available in multiple pack sizes up to 1 kg.
  • Well-characterized identity and physical properties.
Catalog No. 50-003-03243