Learn More
Medchemexpress LLC 2-Chloro-4-iodopyridine | 153034-86-7 | 100.0% | 239.44 | 25 G

Supplier: Medchemexpress LLC 25GHY32754
2-Chloro-4-iodopyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.