Learn More
Medchemexpress LLC 2-Chloro-4-methylphenylboronic acid | 145349-62-8 | 170.40 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3291510G
Encompass_Preferred
2-Chloro-4-methylphenylboronic acid is intended for laboratory chemical use and for the manufacture of substances. It is stable under recommended storage conditions. The most important known symptoms and effects are described in the labeling.
  • Harmful if swallowed.
  • Causes serious eye irritation.
  • Avoid breathing dust, fume, gas, mist, vapours, or spray.
  • Wash hands thoroughly after handling.
  • Do not eat, drink, or smoke when using this product.
  • Use only outdoors or in a well-ventilated area.
  • Wear protective gloves, clothing, and eye/face protection.
  • Store in a cool, well-ventilated area, away from direct sunlight and ignition sources.
Catalog No. 50-004-01664