Learn More
Medchemexpress LLC 2-chloro-4-nitrophenyl beta-D-glucopyranoside | 120221-14-9 | MFCD00037475 | 99.7% | 335.69 g·mol⁻¹ | C12H14ClNO8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0398591G
Encompass_Preferred
2-Chloro-4-nitrophenyl beta-D-glucopyranoside is a chromogenic substrate used to assay β-glucosidase activity. Enzymatic hydrolysis liberates a 2-chloro-4-nitrophenyl chromophore that can be detected colorimetrically, making the compound useful for biochemical assays and as an additive in bacterial growth media. The reagent is supplied as a solid with specified purity and storage recommendations.
Catalog No. 50-003-01214