Learn More
Medchemexpress LLC 2-chloro-4-nitrophenyl β-D-glucopyranoside | 120221-14-9 | MFCD00037475 | 98.0% | 335.69 g/mol | C12H14ClNO8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW039859100MG
Encompass_Preferred
2-Chloro-4-nitrophenyl β-D-glucopyranoside is a chromogenic substrate for detecting β-glucosidase activity. Upon enzymatic hydrolysis it releases 2-chloro-4-nitrophenol, producing a colored product that can be monitored spectrophotometrically. The compound is identified by CAS 120221-14-9, has molecular formula C12H14ClNO8, and is reported at 98% purity.
  • Chromogenic substrate for β-glucosidase assays.
  • Releases a colored product upon hydrolysis for easy detection.
  • Suitable for spectrophotometric monitoring of enzyme activity.
  • Can be used as a supplement in bacterial growth media for detection assays.
  • High reported purity supports consistent assay performance.
  • Available in multiple pack sizes for laboratory use.
Catalog No. 50-002-99454