Learn More
Medchemexpress LLC 2-chloro-4-nitrophenyl β-D-glucopyranoside | 120221-14-9 | MFCD00037475 | 98.0% | 335.69 g/mol | C12H14ClNO8 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW039859
Encompass_Preferred
2-Chloro-4-nitrophenyl β-D-glucopyranoside is a chromogenic substrate used to detect β-glucosidase activity in biochemical assays. Upon enzymatic hydrolysis it releases a colored 2-chloro-4-nitrophenol, enabling spectrophotometric measurement of enzyme activity. The material is supplied at high purity for research and assay development applications.
  • Chromogenic substrate for β-glucosidase assays.
  • Enzymatic cleavage releases 2-chloro-4-nitrophenol for spectrophotometric detection.
  • High purity suitable for analytical and assay applications.
  • Facilitates quantitative measurement of enzyme activity.
  • Applicable to bacterial growth medium formulations.
Catalog No. 50-003-03504