Learn More
Medchemexpress LLC 2-Chloro-4-nitropyridine | 23056-36-2 | 99.9% | 158.54 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010174
Encompass_Preferred
2-Chloro-4-nitropyridine is a drug intermediate in synthesizing other organic compounds, such as 2-chloro-4-ethoxypyridine and various RET kinase inhibitors.
  • Serves as a drug intermediate in chemical synthesis.
  • Utilized in the synthesis of 2-chloro-4-ethoxypyridine.
  • Applicable in the creation of various RET kinase inhibitors.
Catalog No. 50-003-66655