Learn More
Medchemexpress LLC (2-Chloro-4-pyridinyl)methanol | 100704-10-7 | 143.57 | 10 G

Supplier: Medchemexpress LLC HYW00336510G
(2-Chloro-4-pyridinyl)methanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This high-purity compound is provided in solid form.
- Biochemical reagent for life science applications
- Purity of 99.5%
- Molecular weight: 143.57
- Storage at room temperature for 3 years
- In solvent, stable at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.