Learn More
Medchemexpress LLC 2-Chloro-5-cyanopyridine | 33252-28-7 | 99.94% | C6H3ClN2 | 100 G

Supplier: Medchemexpress LLC HY59197100G
2-Chloro-5-cyanopyridine is a biochemical reagent intended for use as a biological material or organic compound in life science-related research.
- CAS number: 33252-28-7
- Molecular weight: 138.55
- Chemical formula: C6H3ClN2
- Appearance: Solid
- Color: White to light yellow
- Solubility: Soluble in DMSO at 100 mg/mL (721.76 mM)
- Powder storage: Store at -20°C for 3 years or 4°C for 2 years
- Storage in solvent: Store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.