Learn More
Medchemexpress LLC 2-Chloro-5-nitrobenzene sulfonamide | 96-72-0 | 236.64 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00849710G
Encompass_Preferred
2-Chloro-5-nitrobenzene sulfonamide is a biochemical reagent intended for research use only. It appears as a solid with a color ranging from white to off-white. The recommended shipping condition for the continental US is room temperature, with variations possible elsewhere.
  • Intended for research use only
  • Solid appearance, white to off-white color
  • Store at room temperature for 3 years
  • For storage in solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-45532