Learn More
Medchemexpress LLC 2-Chloro-5-nitrobenzene sulfonamide | 96-72-0 | 236.64 | 50 G

Supplier: Medchemexpress LLC HYW00849750G
2-Chloro-5-nitrobenzene sulfonamide is a biochemical reagent intended for research use only. This product is not sold to patients.
- Appearance: Solid
- Color: White to off-white
- Formula: C6H5ClN2O4S
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Store at room temperature for 3 years; in solvent -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.