Learn More
Medchemexpress LLC 2-chloro-6-benzylamino-9-methylpurine | 101622-53-1 | MFCD00467327 | 98.2% | 273.72 g·mol⁻1 | C13H12ClN5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW47810225MG
Encompass_Preferred
CDK2-IN-13 is a small-molecule inhibitor of cyclin-dependent kinase 2 (CDK2) used for research into cell cycle regulation and cancer biology. It is reported to have an IC50 of 12 μM. Chemical identifiers include CAS 101622-53-1, molecular formula C13H12ClN5, and molecular weight 273.72 g·mol⁻1. The compound is supplied in milligram quantities for laboratory research.
  • CDK2 inhibition (IC50 12 μM) suitable for target validation studies.
  • Supports cancer and cell cycle research applications.
  • Well-characterized identity (CAS 101622-53-1; C13H12ClN5; MW 273.72).
  • High reported purity for research use (98.2%).
  • Available in small-mass packaging convenient for assay development.
Catalog No. 50-002-84132